Ethyl 1-bromo-2-naphthoate ≥98%

Supplier: Ambeed
Warning

A1146422-10G A1146422-1G A1146422-5G
77865-895EA 60 USD
77865-895 77865-896 77865-897
Ethyl 1-bromo-2-naphthoate ≥98%
Ethyl 1-bromo-2-naphthoate

Precaución: for Research Use Only

Formula: C₁₃H₁₁BrO₂
MDL Number: MFCD06204395
CAS Number: 773134-84-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR