Fmoc-Thr(PO₃H₂)-OH ≥97%

Supplier: Ambeed
Warning

A1146254-1G A1146254-250MG
77783-305EA 225 USD
77783-305 77783-306
Fmoc-Thr(PO₃H₂)-OH ≥97%
Fmoc-Thr(PO₃H₂)-OH

Precaución: for Research Use Only

Formula: C₁₉H₂₀NO₈P
MDL Number: MFCD02684511
CAS Number: 883726-90-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR