1-(2-(Diethylamino)ethyl)-3-(2-(4-ethylpiperazin-1-yl)-4-methylquinolin-6-yl)thiourea ≥98%

Supplier: Ambeed
Danger

Synonyms: D-I03

A1145758-10MG A1145758-1MG A1145758-5MG
77595-808EA 156.83 USD
77595-808 77595-810 77595-812
1-(2-(Diethylamino)ethyl)-3-(2-(4-ethylpiperazin-1-yl)-4-methylquinolin-6-yl)thiourea ≥98%
1-(2-(Diethylamino)ethyl)-3-(2-(4-ethylpiperazin-1-yl)-4-methylquinolin-6-yl)thiourea

Precaución: for Research Use Only

Formula: C₂₃H₃₆N₆S
MW: 428.64 g/mol
MDL Number: MFCD14739485
CAS Number: 688342-78-1
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR