Cilofexor ≥99%

Supplier: Ambeed
Warning

A1145754-1MG A1145754-5MG A1145754-10MG A1145754-100MG A1145754-25MG A1145754-50MG
77595-804EA 32.8 USD
77595-804 77595-806 77595-802 77865-892 77865-893 77865-894
Cilofexor ≥99%
Cilofexor

Precaución: for Research Use Only

Formula: C₂₈H₂₂Cl₃N₃O₅
MDL Number: MFCD31731099
CAS Number: 1418274-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR