(R)-5-Ethyl-3-(6-(3-methoxy-4-methylphenoxy)pyridin-3-yl)imidazolidine-2,4-dione ≥98%

Supplier: Ambeed
Warning

Synonyms: AUT1

A1145734-10MG A1145734-1MG A1145734-5MG
77595-796EA 107.63 USD
77595-796 77595-798 77595-800
(R)-5-Ethyl-3-(6-(3-methoxy-4-methylphenoxy)pyridin-3-yl)imidazolidine-2,4-dione ≥98%
(R)-5-Ethyl-3-(6-(3-methoxy-4-methylphenoxy)pyridin-3-yl)imidazolidine-2,4-dione

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₃O₄
MW: 341.36 g/mol
CAS Number: 1311136-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR