VU0155069 ≥98%

Supplier: Ambeed
Warning

Synonyms: (S)-N-(1-(4-(5-Chloro-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)piperidin-1-yl)propan-2-yl)-2-naphthamide

A1145652-1MG A1145652-5MG
77592-226EA 39.98 USD
77592-226 77595-794
VU0155069 ≥98%
VU0155069

Precaución: for Research Use Only

Formula: C₂₆H₂₇ClN₄O₂
MW: 462.970 g/mol
MDL Number: MFCD23160355
CAS Number: 1130067-06-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR