1,3,5,6-Tetrahydroxy-2-methylanthracene-9,10-dione ≥97%

Supplier: Ambeed
Warning

A1145388-1MG A1145388-10MG A1145388-5MG
77595-792EA 54.33 USD
77595-792 77865-884 77865-885
1,3,5,6-Tetrahydroxy-2-methylanthracene-9,10-dione ≥97%
1,3,5,6-Tetrahydroxy-2-methylanthracene-9,10-dione

Precaución: for Research Use Only

Formula: C₁₅H₁₀O₆
MDL Number: MFCD18448254
CAS Number: 80368-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR