1-Propyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Propyl-1H-pyrazole-5-boronic acid pinacol ester

A1145126-1G A1145126-250MG A1145126-5G
77570-830EA 40 USD
77570-830 77865-876 77865-877
1-Propyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98%
1-Propyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₂H₂₁BN₂O₂
MDL Number: MFCD12405508
CAS Number: 847818-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR