4-Bromo-1-(cyclopropylmethyl)-1H-pyrazole ≥96%

Supplier: Ambeed
Warning

A1145112-5G A1145112-1G A1145112-250MG
77619-978EA 211.15 USD
77619-978 77783-298 77783-299
4-Bromo-1-(cyclopropylmethyl)-1H-pyrazole ≥96%
4-Bromo-1-(cyclopropylmethyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD12403651
CAS Number: 1216152-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR