2,3-Dihydrobenzofuran-7-boronic acid, pinacol ester ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-(2,3-Dihydrobenzofuran-7-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

A1145098-250MG A1145098-100MG A1145098-1G
77595-786EA 59.45 USD
77595-786 77865-872 77865-873
2,3-Dihydrobenzofuran-7-boronic acid, pinacol ester ≥98%
2,3-Dihydrobenzofuran-7-boronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₉BO₃
MDL Number: MFCD11109314
CAS Number: 934586-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR