(2-Ethynylphenyl)boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A1145028-1G
A1145028-100MG
A1145028-250MG
A1145028-5G
77570-826EA
61
USD
77570-826
77865-863
77865-864
77865-865
(2-Ethynylphenyl)boronic acid ≥98%
(2-Ethynylphenyl)boronic acid
Precaución: for Research Use Only
Formula:
C₈H₇BO₂ |
MDL Number:
MFCD10697431 CAS Number: 905926-85-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...