(5-(((tert-Butoxycarbonyl)amino)methyl)thiophen-2-yl)boronic acid ≥98%
Supplier: Ambeed
|
Warning
|
A1145022-1G
A1145022-5G
77570-824EA
35
USD
77570-824
77865-861
(5-(((tert-Butoxycarbonyl)amino)methyl)thiophen-2-yl)boronic acid ≥98%
(5-(((tert-Butoxycarbonyl)amino)methyl)thiophen-2-yl)boronic acid
Precaución: for Research Use Only
Formula:
C₁₀H₁₆BNO₄S |
MDL Number:
MFCD11504845 CAS Number: 1072951-39-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...