4-Formyl-3-methoxyphenylboronic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: (4-Formyl-3-methoxyphenyl)boronic acid

A1145020-1G A1145020-250MG
77619-964EA 74.16 USD
77619-964 77619-966
4-Formyl-3-methoxyphenylboronic acid ≥98%
4-Formyl-3-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MW: 179.965 g/mol
MDL Number: MFCD10697427
CAS Number: 815620-00-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR