Tetrabutylammonium bis-trifluoromethanesulfonimidate ≥97%

Supplier: Ambeed
Danger

Synonyms: Tetrabutylammonium bis((trifluoromethyl)sulphonyl)amide

A1144982-1G A1144982-25G A1144982-5G
77570-818EA 32.96 USD
77570-818 77570-820 77570-822
Tetrabutylammonium bis-trifluoromethanesulfonimidate ≥97%
Tetrabutylammonium bis-trifluoromethanesulfonimidate

Precaución: for Research Use Only

Formula: (CH₃CH₂CH₂CH₂)4N[N(SO₂CF₃)₂
MW: 522.61 g/mol
MDL Number: MFCD01863113
CAS Number: 210230-40-3
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR