tert-Butyl 4-bromo-1H-imidazole-1-carboxylate ≥97%

Supplier: Ambeed
Warning

A1144744-1G A1144744-250MG
77570-816EA 43.26 USD
77570-816 77619-960
tert-Butyl 4-bromo-1H-imidazole-1-carboxylate ≥97%
tert-Butyl 4-bromo-1H-imidazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₈H₁₁BrN₂O₂
MW: 247.09 g/mol
MDL Number: MFCD21364847
CAS Number: 1338257-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR