Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2S)-(+)-1-(4-methoxyphenyl)-1-(4-methoxyphenyl-kC)-3-methyl-1,2-butanediamine]ruthenium(II) ≥98%

Supplier: Ambeed
Danger

A114468-100MG A114468-1G A114468-250MG
77614-896EA 51.5 USD
77614-896 77745-845 77745-846
Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2S)-(+)-1-(4-methoxyphenyl)-1-(4-methoxyphenyl-kC)-3-methyl-1,2-butanediamine]ruthenium(II) ≥98%
Chloro{(S)-(-)-2,2'-bis[di(3,5-xylyl)phosphino]-1,1'-binaphthyl}[(2S)-(+)-1-(4-methoxyphenyl)-1-(4-methoxyphenyl-kC)-3-methyl-1,2-butanediamine]ruthenium(II)

Precaución: for Research Use Only

Formula: C₇₁H₇₃ClN₂O₂P₂Ru
MW: 1184.82 g/mol
MDL Number: MFCD28100836
CAS Number: 1312713-89-5
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR