(R)-1-(5-Fluoro-2-iodophenyl)ethanol ≥98%

Supplier: Ambeed
Warning

A1143702-1G A1143702-25G
77783-293EA 34 USD
77783-293 77783-294
(R)-1-(5-Fluoro-2-iodophenyl)ethanol ≥98%
(R)-1-(5-Fluoro-2-iodophenyl)ethanol

Precaución: for Research Use Only

Formula: C₈H₈FIO
MDL Number: MFCD30802286
CAS Number: 1454852-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR