2-Amino-3-methoxybenzaldehyde ≥97%

Supplier: Ambeed
Warning

A114368-10G A114368-5G
77871-470EA 607 USD
77871-470 77871-471
2-Amino-3-methoxybenzaldehyde ≥97%
2-Amino-3-methoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₉NO₂
MDL Number: MFCD09999245
CAS Number: 70127-96-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR