(2'-Bromo-[1,1'-biphenyl]-2-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

A1143421-5G A1143421-1G
77570-794EA 275.01 USD
77570-794 77619-948
(2'-Bromo-[1,1'-biphenyl]-2-yl)boronic acid ≥97%
(2'-Bromo-[1,1'-biphenyl]-2-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BBrO₂
MW: 276.9200 g/mol
MDL Number: MFCD25973301
CAS Number: 1089189-34-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR