Fmoc-β-HoLys(Boc)-OH ≥98%

Supplier: Ambeed
Warning

A114331-1G A114331-5G A114331-100MG A114331-250MG
77670-598EA 94 USD
77670-598 77670-600 77880-926 77880-927
Fmoc-β-HoLys(Boc)-OH ≥98%
Fmoc-β-HoLys(Boc)-OH

Precaución: for Research Use Only

Formula: C₂₇H₃₄N₂O₆
MW: 482.57 g/mol
MDL Number: MFCD01863054
CAS Number: 203854-47-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR