Pinacidil monohydrate ≥98%

Supplier: Ambeed
Warning

Synonyms: N-Cyano-N′-4-pyridinyl-N′′-(1,2,2-trimethylpropyl)guanidine monohydrate

A1143146-10MG A1143146-5MG
77619-942EA 89.61 USD
77619-942 77619-944
Pinacidil monohydrate ≥98%
Pinacidil monohydrate

Precaución: for Research Use Only

Formula: C₁₃H₁₉N₅·H₂O
MW: 263.3 g/mol
MDL Number: MFCD00242745
CAS Number: 85371-64-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR