13-Bromotridecanoic acid ≥97%

Supplier: Ambeed
Warning

A1143138-250MG A1143138-5G
77592-216EA 75.85 USD
77592-216 77619-940
13-Bromotridecanoic acid ≥97%
13-Bromotridecanoic acid

Precaución: for Research Use Only

Formula: C₁₃H₂₅BrO₂
MW: 293.2404 g/mol
CAS Number: 15462-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR