6-(4-((5-Cyclopropyl-3-(2,6-dichlorophenyl)isoxazol-4-yl)methoxy)piperidin-1-yl)-1-methyl-1H-indole-3-carboxylic acid ≥99%

Supplier: Ambeed
Warning

Synonyms: LY2562175

A1142872-1MG A1142872-10MG A1142872-5MG
77595-770EA 53.3 USD
77595-770 77619-928 77619-930
6-(4-((5-Cyclopropyl-3-(2,6-dichlorophenyl)isoxazol-4-yl)methoxy)piperidin-1-yl)-1-methyl-1H-indole-3-carboxylic acid ≥99%
6-(4-((5-Cyclopropyl-3-(2,6-dichlorophenyl)isoxazol-4-yl)methoxy)piperidin-1-yl)-1-methyl-1H-indole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₂₈H₂₇Cl₂N₃O₄
MW: 540.44 g/mol
MDL Number: MFCD30738721
CAS Number: 1103500-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR