4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid ≥98%

Supplier: Ambeed
Warning

A1142606-100G A1142606-500G
77570-784EA 7.21 USD
77570-784 77570-786
4-Amino-5-hydroxynaphthalene-2,7-disulfonic acid ≥98%
4-Amino-5-hydroxynaphthalene-2,7-disulphonic acid

Precaución: for Research Use Only

Formula: C₁₀H₉NO₇S₂
MW: 319.311 g/mol
MDL Number: MFCD00035728
CAS Number: 90-20-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR