(2S)-2-Acetamido-4-methoxy-4-oxobutanoic acid ≥95%

Supplier: Ambeed
Warning

A1141949-100MG A1141949-1G A1141949-250MG
77865-824EA 31 USD
77865-824 77865-825 77865-826
(2S)-2-Acetamido-4-methoxy-4-oxobutanoic acid ≥95%
(2S)-2-Acetamido-4-methoxy-4-oxobutanoic acid

Precaución: for Research Use Only

Formula: C₇H₁₁NO₅
MW: 189.1659 g/mol
MDL Number: MFCD18326952
CAS Number: 4910-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR