Florfenicol 99+%

Supplier: Ambeed
Danger

Synonyms: 2,2-Dichloro-N-[(1R,2S)-3-fluoro-1-hydroxy-1-(4-methylsulphonylphenyl)-2-propyl]acetamide, D-(-)-threo-2-Dichloroacetamido-3-fluoro-1-(4-methylsulphonylphenyl)-1-propanol, 2,2-Dichloro-N-((1R,2S)-3-fluoro-1-hydroxy-1-(4-(methylsulphonyl)phenyl)propan-2-yl)acetamide

A114157-10G A114157-5G
77599-640EA 16.4 USD
77599-640 77599-642
Florfenicol 99+%
Florfenicol

Precaución: for Research Use Only

Formula: C₁₂H₁₄Cl₂FNO₄S
MW: 358.21 g/mol
MDL Number: MFCD00864834
CAS Number: 73231-34-2
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR