(S)-(1-Methylpyrrolidin-2-yl)methanamine dihydrochloride ≥95%

Supplier: Ambeed
Warning

A114090-250MG A114090-1G A114090-5G
77592-214EA 23.58 USD
77592-214 77610-916 77889-782
(S)-(1-Methylpyrrolidin-2-yl)methanamine dihydrochloride ≥95%
(S)-(1-Methylpyrrolidin-2-yl)methanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₆Cl₂N₂
MDL Number: MFCD27996814
CAS Number: 219320-28-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR