Isoconazole nitrate ≥98%

Supplier: Ambeed
Danger

A113906-5G A113906-25G A113906-1G
77666-654EA 15 USD
77666-654 77666-652 77878-851
Isoconazole nitrate ≥98%
Isoconazole nitrate

Precaución: for Research Use Only

Formula: C₁₈H₁₄Cl₄N₂O
MDL Number: MFCD01721647
CAS Number: 24168-96-5
UN: 1479
ADR: 5.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR