2-Bromo-4-fluoro-5-nitrobenzoic acid ≥98%
Supplier: Ambeed
|
Warning
|
A113904-1G
A113904-5G
77610-912EA
41.2
USD
77610-912
77683-036
2-Bromo-4-fluoro-5-nitrobenzoic acid ≥98%
2-Bromo-4-fluoro-5-nitrobenzoic acid
Precaución: For research use only.
Formula:
C₇H₃BrFNO₄ MW: 264.01 g/mol |
MDL Number:
MFCD11847619 CAS Number: 1036389-83-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...