Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate ≥98%

Supplier: Ambeed
Danger

A113671-100MG A113671-250MG
77614-892EA 119.48 USD
77614-892 77745-843
Bis(1,5-cyclooctadiene)rhodium(I) hexafluoroantimonate ≥98%
Bis(1,5-cyclooctadiene)rhodium (I) hexafluoroantimonate

Precaución: for Research Use Only

Formula: C₁₆H₂₄F₆RhSb
MDL Number: MFCD11113031
CAS Number: 130296-28-5
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR