Givinostat hydrochloride hydrate ≥95%

Supplier: Ambeed
Warning

A113603-100MG A113603-50MG
77685-005EA 319 USD
77685-005 77685-006
Givinostat hydrochloride hydrate ≥95%
Givinostat hydrochloride hydrate

Precaución: for Research Use Only

Formula: C₂₄H₂₇N₃O₄·HCl·H₂O
MDL Number: MFCD18385007
CAS Number: 732302-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR