2-Bromo-5-methylthiazole-4-carboxylic acid ≥97%

Supplier: Ambeed
Warning

A113490-1G A113490-250MG
77670-592EA 47 USD
77670-592 77670-594
2-Bromo-5-methylthiazole-4-carboxylic acid ≥97%
2-Bromo-5-methylthiazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₄BrNO₂S
MW: 222.06 g/mol
MDL Number: MFCD12404925
CAS Number: 1194374-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR