3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 3-cyanobicyclo[1.1.1]pentane-1-carboxylic acid

A113433-250MG A113433-100MG A113433-1G
77570-736EA 207.03 USD
77570-736 77614-884 77747-825
3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid ≥98%
3-Cyanobicyclo[1.1.1]pentane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₇NO₂
MW: 137.136 g/mol
MDL Number: MFCD19229172
CAS Number: 83249-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR