(3-Amino-2,2-difluoropropyl)dimethylamine ≥95%

Supplier: Ambeed
Danger

A1132735-250MG A1132735-1G
77619-896EA 407.88 USD
77619-896 77783-275
(3-Amino-2,2-difluoropropyl)dimethylamine ≥95%
(3-Amino-2,2-difluoropropyl)dimethylamine

Precaución: for Research Use Only

Formula: C₅H₁₂F₂N₂
MDL Number: MFCD28654755
CAS Number: 1601859-07-7
UN: 2734
ADR: 8,3,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR