4-(Trifluoromethyl)phenyl isothiocyanate ≥98%
Supplier: Ambeed
|
Danger
|
A113158-1G
A113158-5G
77670-588EA
10
USD
77670-588
77880-912
4-(Trifluoromethyl)phenyl isothiocyanate ≥98%
4-(Trifluoromethyl)phenyl isothiocyanate
Precaución: for Research Use Only
Formula:
C₈H₄F₃NS |
MDL Number:
MFCD00039645 CAS Number: 1645-65-4 UN: 3261 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...