N1,N6-Bis(2,2,6,6-tetramethylpiperidin-4-yl)hexane-1,6-diamine 4-(4,6-dichloro-1,3,5-triazin-2-yl)morpholine(Poly) MP. 100-140°C

Supplier: Ambeed
Warning

A113109-500G A113109-100G A113109-25G
77570-726EA 47 USD
77570-726 77882-938 77882-939
N1,N6-Bis(2,2,6,6-tetramethylpiperidin-4-yl)hexane-1,6-diamine 4-(4,6-dichloro-1,3,5-triazin-2-yl)morpholine(Poly) MP. 100-140°C
N1,N6-Bis(2,2,6,6-tetramethylpiperidin-4-yl)hexane-1,6-diamine 4-(4,6-dichloro-1,3,5-triazin-2-yl)morpholine(Poly)

Precaución: for Research Use Only

Formula: (C₂₄H₅₀N₄·C₇H₈Cl₂N₄O)x
MDL Number: MFCD00197892
CAS Number: 82451-48-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR