tert-Butyl n-(3-methylpiperidin-3-yl)carbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-methylpiperidin-3-yl)carbamate, 3-Amino-3-methylpiperidine, 3-BOC protected

A113069-1G A113069-5G
77891-816EA 40 USD
77891-816 77891-817
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate ≥95%
tert-Butyl n-(3-methylpiperidin-3-yl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MDL Number: MFCD13194151
CAS Number: 169750-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR