6-Bromo-8-chloroisoquinoline ≥95%

Supplier: Ambeed
Danger

A1129678-1G A1129678-100MG
77570-712EA 541 USD
77570-712 77619-888
6-Bromo-8-chloroisoquinoline ≥95%
6-Bromo-8-chloroisoquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrClN
MW: 242.4997 g/mol
MDL Number: MFCD28129772
CAS Number: 1824270-14-5
UN: 2923
ADR: 8,6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR