2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-8-carboxaldehyde, 1,2-dihydro-5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-1,11-dioxo-, 21-acetate ≥97%

Supplier: Ambeed
Warning

Synonyms: 3-Formyl rifamycin

A112873-5G A112873-1G
77570-708EA 437.75 USD
77570-708 77614-880
2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-8-carboxaldehyde, 1,2-dihydro-5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-1,11-dioxo-, 21-acetate ≥97%
2,7-(Epoxypentadeca[1,11,13]trienimino)naphtho[2,1-b]furan-8-carboxaldehyde, 1,2-dihydro-5,6,9,17,19,21-hexahydroxy-23-methoxy-2,4,12,16,18,20,22-heptamethyl-1,11-dioxo-, 21-acetate

Precaución: for Research Use Only

Formula: C₃₈H₄₇NO₁₃
MW: 725.78 g/mol
MDL Number: MFCD01729454
CAS Number: 13292-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR