3-(3-Fluorophenyl)propiolic acid ≥95%

Supplier: Ambeed
Warning

A1128708-1G A1128708-100MG
77865-810EA 301 USD
77865-810 77865-809
3-(3-Fluorophenyl)propiolic acid ≥95%
3-(3-Fluorophenyl)propiolic acid

Precaución: for Research Use Only

Formula: C₉H₅FO₂
MDL Number: MFCD14707285
CAS Number: 705-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR