5-(tert-Butyl)-2-methoxybenzenecarbaldehyde ≥98%
Supplier: Ambeed
|
Warning
|
A112814-25G
A112814-5G
77599-626EA
317.75
USD
77599-626
77878-849
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde ≥98%
5-(tert-Butyl)-2-methoxybenzenecarbaldehyde
Precaución: for Research Use Only
Formula:
C₁₂H₁₆O₂ |
MDL Number:
MFCD06246086 CAS Number: 85943-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...