1,3,6-Trihydroxy-2-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-9H-xanthen-9-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Neomangiferin

A112790-1MG A112790-10MG A112790-5MG
77610-896EA 12.36 USD
77610-896 77685-000 77685-001
1,3,6-Trihydroxy-2-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-9H-xanthen-9-one ≥98%
1,3,6-Trihydroxy-2-((2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)-7-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-9H-xanthen-9-one

Precaución: for Research Use Only

Formula: C₂₅H₂₈O₁₆
MW: 584.48 g/mol
MDL Number: MFCD04034741
CAS Number: 64809-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR