6-Iodoquinolin-2(1H)-one ≥98%

Supplier: Ambeed
Warning

A112764-1G A112764-5G
77880-906EA 71 USD
77880-906 77880-907
6-Iodoquinolin-2(1H)-one ≥98%
6-Iodoquinolin-2(1H)-one

Precaución: for Research Use Only

Formula: C₉H₆INO
MDL Number: MFCD15144678
CAS Number: 99455-01-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR