2-(2,2-Dimethylbenzo[d][1,3]dioxol-5-yl)acetic acid ≥95%

Supplier: Ambeed
Warning

A1127503-100MG A1127503-1G
77619-882EA 153.47 USD
77619-882 77619-884
2-(2,2-Dimethylbenzo[d][1,3]dioxol-5-yl)acetic acid ≥95%
2-(2,2-Dimethylbenzo[d][1,3]dioxol-5-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₄
MW: 208.2106 g/mol
MDL Number: MFCD24485341
CAS Number: 38515-59-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR