4-(Azetidin-1-yl)benzaldehyde ≥95%

Supplier: Ambeed
Warning

A1126469-100MG A1126469-1G
77783-260EA 40 USD
77783-260 77783-261
4-(Azetidin-1-yl)benzaldehyde ≥95%
4-(Azetidin-1-yl)benzaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO
MDL Number: MFCD24684251
CAS Number: 353247-81-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR