3-Bromo-1,1-difluorocyclobutane ≥98%
Supplier: Ambeed
|
Warning
|
A112575-100MG
A112575-250MG
77891-811EA
35
USD
77891-811
77891-812
3-Bromo-1,1-difluorocyclobutane ≥98%
3-Bromo-1,1-difluorocyclobutane
Precaución: for Research Use Only
Formula:
C₄H₅BrF₂ |
MDL Number:
MFCD22989493 CAS Number: 1310729-91-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...