Y-27632 ≥98%

Supplier: Ambeed
Warning

Synonyms: trans-4-((R)-1-Aminoethyl)-N-(pyridin-4-yl)cyclohexanecarboxamide

A112358-1MG A112358-5MG A112358-10MG
77599-622EA 15.38 USD
77599-622 77599-624 77599-620
Y-27632 ≥98%
Y-27632

Precaución: for Research Use Only

Formula: C₁₄H₂₁N₃O
MDL Number: MFCD03093855
CAS Number: 146986-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR