(3bR,9bS)-6-Hydroxy-7-isopropyl-9b-methyl-3b,4,5,9b,10,11-hexahydrophenanthro[1,2-c]furan-1(3H)-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Triptophenolide

A112189-10MG A112189-1MG A112189-5MG
77614-872EA 73.13 USD
77614-872 77614-874 77614-876
(3bR,9bS)-6-Hydroxy-7-isopropyl-9b-methyl-3b,4,5,9b,10,11-hexahydrophenanthro[1,2-c]furan-1(3H)-one ≥98%
(3bR,9bS)-6-Hydroxy-7-isopropyl-9b-methyl-3b,4,5,9b,10,11-hexahydrophenanthro[1,2-c]furan-1(3H)-one

Precaución: for Research Use Only

Formula: C₂₀H₂₄O₃
MW: 312.4 g/mol
MDL Number: MFCD00210567
CAS Number: 74285-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR