2-(Benzyloxy)ethyl 4-methylbenzenesulfonate ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Benzyl-PEG1-Tos
A112142-1G
A112142-250MG
77684-995EA
87
USD
77684-995
77684-996
2-(Benzyloxy)ethyl 4-methylbenzenesulfonate ≥95%
2-(Benzyloxy)ethyl 4-methylbenzenesulfonate
Precaución: for Research Use Only
Formula:
C₁₆H₁₈O₄S MW: 306.37 g/mol |
MDL Number:
MFCD00027610 CAS Number: 4981-83-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...