2-Chloroimidazo[1,2-a]pyridine-3-carbonitrile ≥95%

Supplier: Ambeed
Warning

A1118871-250MG A1118871-5G
77619-872EA 80.34 USD
77619-872 77783-249
2-Chloroimidazo[1,2-a]pyridine-3-carbonitrile ≥95%
2-Chloroimidazo[1,2-a]pyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₈H₄ClN₃
MDL Number: MFCD09041343
CAS Number: 131773-48-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR